Anti-CDKN1C Mouse Monoclonal Antibody (Purified, BSA-free) [clone: KIP2/880]

Leverandør: Biotium
BNUM0880-50
BNUM0880-50EA 5125 NOK
BNUM0880-50
Anti-CDKN1C Mouse Monoclonal Antibody (Purified, BSA-free) [clone: KIP2/880]
Antistoffer
Recognizes a protein of 57 kDa, identified as p57Kip2. It shows no cross-reaction with p27Kip1. p57Kip2 is a potent tight-binding inhibitor of several G1 cyclin complexes, and is a negative regulator of cell proliferation. Anti-p57 has been used as an aide in identification of complete hydatidiform mole (CHM) (no nuclear labeling of cytotrophoblasts and stromal cells) from partial hydatidiform mole (PHM) in which both cytotrophoblasts and stromal cells stain. The histological differentiation of complete mole, partial mole, and hydropic spontaneous abortion is problematic. Most completeMore Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol CDKN1C
Antigen-navn Cyclin-dependent kinase inhibitor 1C
Environmentally Preferable
Antigen synonymer BWCR|p57|Cyclin dependent kinase inhibitor 1C (CDKN1C)|Beckwith Wiedemann syndrome (WBS)|p57Kip2|KIP2|Cyclin dependent kinase inhibitor p57
Antistofftype Primary
Klonalitet Monoclonal
Konjugering Unconjugated
Klon Clone KIP2/880
Reaktivitet Human,Mouse
Vert Mouse
Gen-ID 106070
Isotype IgG2b kappa
ImmunoKjemi Yes
ImmunoFluorescens Yes
Rensing BSA-free
Konsentrasjon 1 mg/mL
Lagringsbuffer PBS/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR