Anti-BrdU Mouse Monoclonal Antibody (Purified, BSA-free) [clone: BRD.3]

Leverandør: Biotium
BNUM0885-50
BNUM0885-50EA 5125 NOK
BNUM0885-50
Anti-BrdU Mouse Monoclonal Antibody (Purified, BSA-free) [clone: BRD.3]
Antistoffer
This antibody reacts with Bromodeoxyuridine (BrdU) in single stranded DNA (produced by partial denaturation of double stranded DNA), BrdU coupled to a protein carrier, as well as free BrdU. BrdU is a thymidine analog, incorporated into cell nuclei during DNA synthesis prior to mitosis. Antibody to BrdU is helpful in detecting S-phase cells, providing useful information on the aggressiveness of tumors.

Type: Primary
Antigen: Not Applicable
Clonality: Monoclonal
Clone: BRD.3
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: All species
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR