Anti-MUC5AC Mouse Monoclonal Antibody (Purified, BSA-free) [clone: 58M1]
Leverandør: Biotium
BNUM1003-50
BNUM1003-50EA 5125 NOK
BNUM1003-50
Anti-MUC5AC Mouse Monoclonal Antibody (Purified, BSA-free) [clone: 58M1]
Antistoffer
This MAb recognizes the peptide core of gastric mucin M1 (recently identified as Mucin 5AC). Its epitope is destroyed by beta-mercaptoethanol but not by periodate treatment. This mucin is present in primary ovarian mucinous cancer but usually absent in colorectal adenocarcinoma, thus showing an expression pattern opposite to MUC2. Together with a panel of antibodies, Anti-MUC5AC may be useful for differential identification of primary mucinous ovarian tumors from colon adenocarcinoma metastatic to the ovary. MUC5AC antibodies may also be useful for identification of intestinal metaplasia asMore Product Information