Anti-ITGA2B Mouse Monoclonal Antibody (Purified, BSA-free) [clone: ITGA2B/1036]

Leverandør: Biotium
BNUM1036-50
BNUM1036-50EA 5125 NOK
BNUM1036-50
Anti-ITGA2B Mouse Monoclonal Antibody (Purified, BSA-free) [clone: ITGA2B/1036]
Antistoffer
Reacts with a calcium-dependent complex of CD41/CD61, a dimer of 90 kDa and 140 kDa present on the membrane of normal platelets and megakaryocytes. CD41/CD61 is also known as platelet glycoprotein GPIIb/GPIIIa or integrin IIa/3. This complex is the receptor of fibrinogen, fibronectin and von Willebrand factor, and mediates platelet adhesion and aggregation.

Type: Primary
Antigen: ITGA2B
Clonality: Monoclonal
Clone: ITGA2B/1036
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR