Anti-HLA-A; HLA-B; HLA-C Mouse Monoclonal Antibody (Purified, BSA-free) [clone: 246-B8.E7]

Leverandør: Biotium
BNUM1054-50
BNUM1054-50EA 5125 NOK
BNUM1054-50
Anti-HLA-A; HLA-B; HLA-C Mouse Monoclonal Antibody (Purified, BSA-free) [clone: 246-B8.E7]
Antistoffer
Reacts with a monomorphic determinant of human major histocompatibility (MHC) class I antigens (HLA-A, B and C). Human MHC class I antigens are expressed constitutively on all nucleated cells lymphocytes such as lymphocytes, thymocytes, granulocytes, and bone marrow cells and are absent on erythrocytes. MHC class I antigens play a role in class I MHC-associated antigen presentation, inhibition of NK cell cytotoxicity, tumor surveillance, and tissue allotransplantation.

Type: Primary
Antigen: HLA-A; HLA-B; HLA-C
Clonality: Monoclonal
Clone: 246-B8.E7
Conjugation: Purified, BSA-free
Epitope:
More Product Information
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR