Wire baskets, rectangular, with handle
Leverandør: BOCHEM
10120
BOCH10120EA 7725 NOK
BOCH10120
Wire baskets, rectangular, with handle
Kurver Trådkurver
These rectangular stainless steel baskets are designed for transporting, storing, cleaning and sterilising laboratory utensils.
  • Spot-welded grid 8×8×1 mm
  • Reinforced edges
  • Two cross-ties