Transport trolleys
Leverandør: BOCHEM
17330
BOCH17330EA 15575 NOK
BOCH17330
Transport trolleys
Traller
Transport trolley made of electrolytically polished stainless steel.
  • With handle and four guide rolls (two lockable).
Bestillingsinformasjon: Please order baskets separately.