Transport trolleys for baskets, EURO-STANDARD
Leverandør: BOCHEM
17333
BOCH17333EA 28950 NOK
BOCH17333
Transport trolleys for baskets, EURO-STANDARD
Traller
Transport cart made of stainless steel used for transport baskets.
  • Five hanging ties
  • Two pushing handles
  • Two guide and fixed rolls, two with lock
  • Enclosure of rolls, zincked
  • Excess length of enclosure