Burette stand, DUPLEX
Leverandør: BOCHEM
5620
BOCH5620EA 2650 NOK
BOCH5620
Burette stand, DUPLEX
Stativer
This burette stand is made of green varnished cast iron.
  • Incl. stainless steel rod (600×10 mm)
  • Incl. two plates with diameter 105 mm (black and white)
  • For use with burette holder (article no. 5600)