Microplate shakers, Jitterbug

Leverandør: BOEKEL SCIENTIFIC
130000-2
BOEI130000-2EA 0 NOK
BOEI130000-2
Microplate shakers, Jitterbug
Risteapparater Risteapparater, mikroplater
The Jitterbug microplate shaker is ideal for immunoassays like ELISA, PCR and other research procedures requiring heating and shaking of microplates.

  • Accommodates two standard microplates (96-, 384-, 1536-well) or strip trays
  • Independently set temperature, shaking speed and mixing duration
  • Select from eight mixing speeds
  • Smooth, quiet operation
  • Excellent temperature uniformity

Includes inner lid to reduce the risk of contamination.
Bestill nå

SPESIFIKASJONER

Modell Jitterbug
Environmentally Preferable
Hastighetsområde 575 − 1500 min⁻¹
kretsløp 1,0 mm
Antall mikroplater det er plass til 2
Ristebevegelse Rotating counterweight
Vekt 4,99 kg
B×D×H 254×254×102 mm
Elektronisk 230 V, 50/60 Hz

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR