Adapters, receiver, BOLA
Leverandør: Bohlender
B292-02
BOHLB292-02EA 3550 NOK
BOHLB292-02
Adapters, receiver, BOLA
Destillasjonsutstyr Destillasjonsadaptere
PTFE, white.
  • Temperature resistance: from −200 to +250 °C
  • Universal chemical resistance
Ground joint cone and socket size NS 29, as well as socket size NS 19 for example for vacuum stopcock (see Cat. No. B 293-02). Integrated special nut (Safe-Lab) for easy locking and unlocking of the ground joint.