Vacuum stopcocks, BOLA
Leverandør: Bohlender
B293-02
BOHLB293-02EA 6825 NOK
BOHLB293-02
Vacuum stopcocks, BOLA
Haner
Made of PTFE.
  • Universal chemical resistance
  • Temperature resistance: from 0 to +110 °C
Vacuum stopcock with ground joint cone, bore and connections for tubing. Integrated special nut (Safe-Lab) for easy locking and unlocking of the ground joint.