Distilling links, BOLA
Leverandør: Bohlender
B303-02
BOHLB303-02EA 4850 NOK
BOHLB303-02
Distilling links, BOLA
Destillasjonsutstyr Destillasjonsadaptere
PTFE, white.
  • Temperature resistance: from −200 to +250 °C
  • Universal chemical resistance
  • Autoclavable
For vertical positioning of the Liebig condenser (e.g. in a lid of a reaction vessel). Integrated special nut (Safe-Lab) for easy locking and unlocking of the ground joint.