Cold traps, BOLA
Leverandør: Bohlender
B317-60
BOHLB317-60EA 24225 NOK
BOHLB317-60
Cold traps, BOLA
Vacuumfeller
PTFE head with two threaded necks GL14 for connection of hard-walled tubing by means of separately available laboratory screw joints. Connecting nut is made of PTFE reinforced with glassfibres. GL hose connectors are recommend for soft, flexible tubing. Temperature resistant from −200 to +250 °C with suitable vaccum transparent. PFA, transparent.