Multiple distributors with ground joints, BOLA
Leverandør: Bohlender
D620-08
BOHLD620-08EA 2050 NOK
BOHLD620-08
Multiple distributors with ground joints, BOLA
Adaptere Slangeskjøter med NS-slip
PTFE, white. For bottles or reaction vessels with ground joint. For transferring liquids without contamination. For connection to a liquid source or a pressure or vacuum system.
  • Temperature resistance: −200 to +250 °C
  • Universal chemical resistance
  • Autoclavable
  • Vacuum: suitable
Completely made of PTFE. With ground joint size 29/32 and two GL-threaded necks. Integrated special nut for easy unlocking of the ground joint. For the connection of tubing or tube made of glass, plastic or metal by means of laboratory screw joints.