Multiple distributors for bottles, EX, BOLA
Leverandør: Bohlender
D864-08 D865-08
BOHLD864-08EA 2200 NOK
BOHLD864-08 BOHLD865-08
Multiple distributors for bottles, EX, BOLA
Lokk Flaskelokk
Black screw cap, made of conductive PPS-EX, for GL 45 threaded bottles and distributor body made of conductive PTFE-EX.
  • Very good chemical resistance
  • Temperature resistant from −20 to +200 °C
  • Suitable for vacuum applications
For the removal or filling of aggressive or pure liquids. For the connection of hoses, tubes and probes into vessels. Hoses with a maximum external diameter of  8,5 mm can be inserted through the necks.