Adapters for chromatography applications, BOLA
Leverandør: Bohlender
F757-03 F757-09 F755-03 F755-15 F757-15 F757-12 F755-09 F757-06 F755-12 F755-06
BOHLF757-03EA 495 NOK
BOHLF757-03 BOHLF757-09 BOHLF755-03 BOHLF755-15 BOHLF757-15 BOHLF757-12 BOHLF755-09 BOHLF757-06 BOHLF755-12 BOHLF755-06
Adapters for chromatography applications, BOLA
Fittings og kontakter
Black, screw cap made of PPS with GL thread and body made of PTFE with female thread UNF 1/4” 28G. The bottle neck is adjusted by an o-ring behind an elastic sealing lip.
  • Temperature resistance: from −50 to 200 °C
  • Very good chemical resistance
  • FDA conformity