Basket dipper, BOLA
Leverandør: Bohlender
H997-03
BOHLH997-03EA 2200 NOK
BOHLH997-03
Basket dipper, BOLA
Prøvetakere Prøveøser
Non porous basket dipper designed for washing, rinsing or dipping solids.
  • Good temperature resistance from −200 to +250 °C
  • Excellent chemical resistance
  • Mesh in the bottom (1,0x1,0 mm) and a fixed stem
Suitable for use with aggressive or pure substances, such as boiling stones and balls.
Sertifiseringer: FDA conform.