Vacuum adapter, GL thread, PTFE, BOLA
Leverandør: Bohlender
N1656-45
BOHLN1656-45EA 2075 NOK
BOHLN1656-45
Vacuum adapter, GL thread, PTFE, BOLA
Adaptere Slangeskjøter med NS-slip
This vacuum adapter is made of PTFE with O-ring made of FKM for secure placement of standard filter cartridges made of glass or plastic on flasks with GL 45 thread.
  • Lateral step-type solenoid made of PTFE
  • Chemical resistance
  • Temperature resistance from -15 to +200 °C
For vacuum filtration, the O-ring at the passageway seals the connection to the filter slide so that even a small vacuum is sufficient.