Total immersion temperature probes, BOLA
Leverandør: Bohlender
P1780-20
BOHLP1780-20EA 3850 NOK
BOHLP1780-20
Total immersion temperature probes, BOLA
Temperaturprober
Total immersion PTFE probe (class A, PRT 100), with white PFA-coated cable (4 strands) and collar ring Ø 5 mm.
  • Typical response times of T 50 is 4 to 6 seconds and T 90 is 6 to 8 seconds
Used for measuring temperatures in autoclaves and can be immersed totally into the liquid.
Sertifiseringer: FDA confirmed.