Magnet holder for waste pipes, b.safe
Leverandør: Bohlender
R1162-32
BOHLR1162-32EA 550 NOK
BOHLR1162-32
Magnet holder for waste pipes, b.safe
Veggkrok
Pipe clamp made of PE with permanently fixed power magnets. b.safe Waste Pipes can easily be clipped into the pipe clamp for tube Ext.Ø 32 mm.
  • Compact collective connector to combine the outlets of two respectively up to four HPLC units
  • All connectors are placed sideways to join tubing and tubes with suitable fittings and hose connectors without bending
In combination with a suitable desktop- or wall mount, b.safe Waste Pipes can easily be fixed in height by means of the magnet.