Mount for waste pipes, b.safe
Leverandør: Bohlender
R1163-32
BOHLR1163-32EA 550 NOK
BOHLR1163-32
Mount for waste pipes, b.safe
Kromatografiutstyr Kromatografireservedeler
Set consisting of (1) base plate with female thread M8, (1) threaded rod M8, length 200 mm, (1) pipe clamp with female thread M8 for tube ext.Ø 32 mm, and fastening material for assembly on walls or under desktops.
  • Base plate dimensions: 80×30×3 mm
Fix the base plate on the wall. First connect the threaded rod with the pipe clamp, then connect the pipe clamp to the base plate. Make sure to align the clamp correctly. Open the clamp, clip in the b.safe Waste Pipe and close the clamp again.