Wall mounting bracket for waste pipes, b.safe
Leverandør: Bohlender
R1164-03
BOHLR1164-03EA 470 NOK
BOHLR1164-03
Wall mounting bracket for waste pipes, b.safe
Veggkrok
Black, powder-coated plate made of steel. With 2 bores Ø 4,2 mm for wall mounting with the included fixing material.
  • Compact collective connector to combine the outlets of two respectively up to four HPLC units
  • All connectors are placed sideways to join tubing and tubes with suitable fittings and hose connectors without bending
Waste Pipes with tube Ext.Ø 32 mm can be fixed and adjusted in height on the long side of the plate by using suitable b.safe Magnet Holders.