Desktop mounting bracket for waste pipes, b.safe
Leverandør: Bohlender
R1165-03
BOHLR1165-03EA 405 NOK
BOHLR1165-03
Desktop mounting bracket for waste pipes, b.safe
Veggkrok
Contact area with silicone-free glue pad and 2 bores Ø 4,2 mm for non-slip fixing on the desktop.
  • Black, powder-coated angle plate made of steel
  • Compact collective connector to combine the outlets of two respectively up to four HPLC units
  • All connectors are placed sideways to join tubing and tubes with suitable fittings and hose connectors without bending
Waste Pipes with tube Ext.Ø 32 mm can be fixed and adjusted in height on the long side of the plate by using suitable b.safe Magnet Holders.