Direct filling system, b.safe
Leverandør: Bohlender
R1500-05
BOHLR1500-05EA 146600 NOK
BOHLR1500-05
Direct filling system, b.safe
Sikkerhetskanner
Highly specialised system for the disposal of especially viscous liquids or liquid waste with solid contents.
  • Telescopic disposal tube made of stainless steel
  • All parts are suitable for use in explosion-proof environments and prepared for installation on site
  • For the safe disposal of all kinds of liquid waste
Only a b.safe Funnel with hinged lid and the b.safe Mini Display for controlling the filling level of your waste canister are visible in the working area of your hood.
Leveringsinformasjon: Includes:
- b.safe Funnel 130 made of staticMore Product Information