Isolated amplifier, b.safe
Leverandør: Bohlender
R660-01
BOHLR660-01EA 11450 NOK
BOHLR660-01
Isolated amplifier, b.safe
Nivåsensorer
Enclosed housing for installation at the rear side of a safety cabinet. Unit provides safe signal exchange between explosion-hazardous and explosion-proof areas by galvanic isolation of the sensor side and the output side.
  • For galvanic isolation of the sensor side and the output side
  • Ignitable sparks are prevented in the explosive atmosphere on the sensor side
Ready for installation plastic housing made of ABS (IP 66 rated) that prevents accidental contact of live parts. With clearly labelled housing screw connections to connect up to two b.safeMore Product Information