b.safe Isolated Amplifier 1-Channel
Leverandør: Bohlender
R660-04
BOHLR660-04EA 8775 NOK
BOHLR660-04
b.safe Isolated Amplifier 1-Channel
Dataloggere
Ready-to-install plastic housing made of ABS (IP66) protects against accidental contact with live contacts.
  • With clearly labelled housing screw connections to connect b.safe filling level sensors
  • 1-channel version for use with one approved level sensor
  • 2-channel for two approved level sensors and their safe operation in explosion-proof areas
Encapsulated housing for an installation, such as at the back wall of a safety cabinet. Safe signal exchange between explosion-hazardous and explosion-proof areas by galvanic isolation of the sensor side and the
More Product Information
New Product
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Grensesnitt