Antistatic corrugated tubing, b.safe
Leverandør: Bohlender
R773-16 R773-18
BOHLR773-16EA 3275 NOK
BOHLR773-16 BOHLR773-18
Antistatic corrugated tubing, b.safe
Slanger Overføringsslange
Flexible corrugated tubing made of static dissipative plastics.
Ordered quantities are supplied in continuous rolls by the metre.