Starter set, b.safe Multiport IV
Leverandør: Bohlender
R9125-04
BOHLR9125-04EA 5400 NOK
BOHLR9125-04
Starter set, b.safe Multiport IV
Fittings og kontakter
One connection for collecting liquid waste from up to 4 HPLC systems that can be integrated directly in laboratory hoods or working tables. b.safe Waste Tubes lead liquid solvent waste directly into connected canisters.
  • Compact collective connector to combine the outlets of two respectively up to four HPLC units
  • All connectors are placed sideways
  • Additional slope improves the flow and prevents any backflow
Made of static dissipative PE-EX, suitable for boards with a thickness of 10 to 35 mm, easy fixing with counter nut. Low installation height due toMore Product Information