Virginal rods, BOLA
Leverandør: Bohlender
S1800-06 S1800-12 S1800-25 S1800-30 S1800-40 S1800-20 S1800-16 S1800-15 S1800-10 S1800-08
BOHLS1800-06EA 605 NOK
BOHLS1800-06 BOHLS1800-12 BOHLS1800-25 BOHLS1800-30 BOHLS1800-40 BOHLS1800-20 BOHLS1800-16 BOHLS1800-15 BOHLS1800-10 BOHLS1800-08
Virginal rods, BOLA
Stativstang
Virginal rods made of white PTFE.
  • Diameter and length are nominal dimensions and can contain a machining allowance
  • Temperature resistance from −200 to +250 °C
  • Universal chemical resistance
  • FDA conform