Surface protection tiles, BOLA
Leverandør: Bohlender
S1805-06 S1805-02 S1805-08 S1805-16 S1805-14 S1805-12 S1805-10 S1805-04
BOHLS1805-06EA 1800 NOK
BOHLS1805-06 BOHLS1805-02 BOHLS1805-08 BOHLS1805-16 BOHLS1805-14 BOHLS1805-12 BOHLS1805-10 BOHLS1805-04
Surface protection tiles, BOLA
Benkepapir Benkepapir
For use as a table pad, slideway or for insulation.
  • Protection mats made from PTFE
  • Standard sizes with different thicknesses
  • Temperature resistant from –200 to +250 °C