Maintenaince Kit for Star Desiccators
Leverandør: Bohlender
V1778-02
BOHLV1778-02EA 1250 NOK
BOHLV1778-02
Maintenaince Kit for Star Desiccators
Eksikatorer
Maintenance Kit for SICCO Star-Desiccators consists of one cellular rubber sealing tape, one magnetic tape and four non-slip rubber feet.
  • Easy replacement of sealing and magnetic tape to ensure the airtight closure of the desiccator