Desiccant, SICCO
Leverandør: Bohlender
V1895-12
BOHLV1895-12EA 5325 NOK
BOHLV1895-12
Desiccant, SICCO
Eksikatorer Tørkemidler
Silica gel with colour indicator.
  • Does not contain cobalt-II-chloride
  • Grain size: 2,0 - 5,0 mm
  • Loss after drying ≤4%
Colour indicator changes from orange to dark brown and black.