Desiccator cabinet, anti-static, SICCO Star
Leverandør: Bohlender
V1910-07
BOHLV1910-07EA 45500 NOK
BOHLV1910-07
Desiccator cabinet, anti-static, SICCO Star
Eksikatorer Eksikatorskap
PC desiccator cabinet which has reinforced aluminium frame with antistatic acrylic panels and easily readable electronic hygrometer. Mainly used for temporary storage of components for electronic products.
  • Variable height telescopic shelves made of aluminium or stainless steel
  • Non-slip rubber feet or four casters (two of the casters with brakes)
  • Door with magnetic catch and circular rubber seal
  • Connection for grounding cable and tray for desiccant
These desiccators can protect electronic components against humidity and airborne particulate.More Product Information