Desiccator cabinets, anti-static, SICCO maxi series
Leverandør: Bohlender
V1922-07 V1923-07
BOHLV1922-07EA 98150 NOK
BOHLV1922-07 BOHLV1923-07
Desiccator cabinets, anti-static, SICCO maxi series
Eksikatorer Eksikatorskap
Desiccator cabinets, PC ESD, have a reinforced aluminium frame with panels made of antistatic polycarbonate and an easy to read electronic hygrometer. They are mainly used for temporary storage of components for electronic products.
  • Door with magnetic catch and circular rubber seal
  • Four casters (two of the casters with brakes)
  • Variable height telescopic shelves made of stainless steel
  • Connection for grounding cable and tray for desiccant
These desiccators can protect electronic components against humidity and airborne particulate. ElectrostaticMore Product Information