Glove box, anti-static, with transfer chamber, BOLA
Leverandør: Bohlender
V1992-08
BOHLV1992-08EA 0 NOK
BOHLV1992-08
Glove box, anti-static, with transfer chamber, BOLA
Hanskebokser
This anti-static glove box made of PC-ESD allows safe working with sensitive compounds in a controlled environment.
  • Robust and lightweight aluminium construction
  • Superb all-round visibility
  • Non slip rubber feet
  • Cable lead-ins for electric devices
  • Large front opening
  • Connectors for gas-filling and aeration including appropriate tubing
  • Transfer chamber for inserting further products
Aluminium frame with static dissipative panels made of polycarbonate, base panel made of static dissipative PE. Front panel with two glove ports and one pair of staticMore Product Information
Bestill nå
SPESIFIKASJONER
Modell V 1192-08
Volum 290 L
Environmentally Preferable
Overføringskammer (B × D × H) (mm) 220×320×275 mm
B × D × H utv. 1210×600×700 mm
B × D × H inv. 850×540×500 mm
Vekt 20 kg