Desiccator cabinet, SICCO wall
Leverandør: Bohlender
V1994-08
BOHLV1994-08EA 236300 NOK
BOHLV1994-08
Desiccator cabinet, SICCO wall
Eksikatorer Eksikatorskap
Transparent PPMA cabinet with reinforced aluminium frame, unit has a door with a foam rubber gasket and magnetic strip. Designed for storing or drying humidity sensitive products using silica gel. The controlled environment inside the desiccator is ideal for storing reference materials, retained samples, metallographic specimens, tobacco and DNA-samples.
  • Door mounted electronic hygrometer indicates minimum and maximum temperature and humidity values
  • Temperature resistance from −20 to +70 °C
  • Easy to move about as unit has four castors, two of them with