Cleanroom goggles, Coverall Autoclave
Leverandør: Bollé Safety
COVACLAVE
BOLECOVACLAVEEA 420 NOK
BOLECOVACLAVE
Cleanroom goggles, Coverall Autoclave
Vernebriller /Goggles
These cleanroom Coverall goggles with interchangeable ventilation ensure protection and comfort. It is possible to wear prescription spectacles under them.
  • PLATINUM coated lens: Coating on both sides ensures high scratch-resistance (1,4 cd/m²), high resistance to the most aggressive chemicals and slows the appearance of fogging
  • Adjustable strap
  • Upper indirect and interchangeable ventilation
  • Ultra-comfort autoclavable goggles
Weight: 54 g
Sertifiseringer: EN 166