Anti-ADM Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Leverandør: Bioss
BS-0007R-A750
BOSSBS-0007R-A750EA 10425 NOK
BOSSBS-0007R-A750
Anti-ADM Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antistoffer
AM and PAMP are potent hypotensive and vasodilatator agents. Numerous actions have been reported most related to the physiologic control of fluid and electrolyte homeostasis. In the kidney, am is diuretic and natriuretic, and both am and pamp inhibit aldosterone secretion by direct adrenal actions. In pituitary gland, both peptides at physiologically relevant doses inhibit basal ACTH secretion. Both peptides appear to act in brain and pituitary gland to facilitate the loss of plasma volume, actions which complement their hypotensive effects in blood vessels.
Type: Primary
Antigen:More Product Information