Anti-IGF2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Leverandør: Bioss
BS-0015R-A350
BOSSBS-0015R-A350EA 10450 NOK
BOSSBS-0015R-A350
Anti-IGF2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antistoffer
The insulin-like growth factors possess growth-promoting activity. In vitro, they are potent mitogens for cultured cells. IGF-II is influenced by placental lactogen and may play a role in fetal development. Preptin undergoes glucose-mediated co-secretion with insulin, and acts as physiological amplifier of glucose-mediated insulin secretion. Exhibits osteogenic properties by increasing osteoblast mitogenic activity through phosphoactivation of MAPK1 and MAPK3.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IGF2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
More Product Information
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR