Anti-IGF2 Rabbit Polyclonal Antibody (Cy7®)

Leverandør: Bioss
BS-0015R-CY7
BOSSBS-0015R-CY7EA 10550 NOK
BOSSBS-0015R-CY7
Anti-IGF2 Rabbit Polyclonal Antibody (Cy7®)
Antistoffer
The insulin-like growth factors possess growth-promoting activity. In vitro, they are potent mitogens for cultured cells. IGF-II is influenced by placental lactogen and may play a role in fetal development. Preptin undergoes glucose-mediated co-secretion with insulin, and acts as physiological amplifier of glucose-mediated insulin secretion. Exhibits osteogenic properties by increasing osteoblast mitogenic activity through phosphoactivation of MAPK1 and MAPK3.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IGF2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host:More Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol IGF2
Environmentally Preferable
Antigen-navn Insulin-like Growth Factor 2
Antigen synonymer IGF-II|PP9974|GRDF|C11orf43
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Cy7®
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 3481
Isotype IgG
ImmunoKjemi Yes
Immunogen 20-70/180
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at 4°C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR