Anti-CREB1 Rabbit Polyclonal Antibody (Cy5®)

Leverandør: Bioss
BS-0036R-CY5
BOSSBS-0036R-CY5EA 10600 NOK
BOSSBS-0036R-CY5
Anti-CREB1 Rabbit Polyclonal Antibody (Cy5®)
Antistoffer
Phosphorylation-dependent transcription factor that stimulates transcription upon binding to the DNA cAMP response element (CRE), a sequence present in many viral and cellular promoters. Transcription activation is enhanced by the TORC coactivators which act independently of Ser-133 phosphorylation. Involved in different cellular processes including the synchronization of circadian rhythmicity and the differentiation of adipose cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CREB1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR