Anti-ADNP Rabbit Polyclonal Antibody (Cy3®)

Leverandør: Bioss
BS-0039R-CY3
BOSSBS-0039R-CY3EA 10350 NOK
BOSSBS-0039R-CY3
Anti-ADNP Rabbit Polyclonal Antibody (Cy3®)
Antistoffer
The activity dependent neuroprotective protein (ADNP) gene is frequently amplified in many neoplasias, including breast, bladder, ovarian, pancreatic, and colon cancers. ADNP mRNA is abundantly expressed in distinct normal tissues, and high expression levels were encountered in malignant cells. ADNP is implicated in maintaining cell survival, perhaps through modulation of p53. The encoded protein contains one homeobox and nine zinc finger domains, suggesting that it functions as a transcription factor.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ADNP
Clonality:More Product Information
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR