Anti-ADNP Rabbit Polyclonal Antibody (Cy5®)

Leverandør: Bioss
BS-0039R-CY5
BOSSBS-0039R-CY5EA 10350 NOK
BOSSBS-0039R-CY5
Anti-ADNP Rabbit Polyclonal Antibody (Cy5®)
Antistoffer
The activity dependent neuroprotective protein (ADNP) gene is frequently amplified in many neoplasias, including breast, bladder, ovarian, pancreatic, and colon cancers. ADNP mRNA is abundantly expressed in distinct normal tissues, and high expression levels were encountered in malignant cells. ADNP is implicated in maintaining cell survival, perhaps through modulation of p53. The encoded protein contains one homeobox and nine zinc finger domains, suggesting that it functions as a transcription factor.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ADNP
Clonality:More Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol ADNP
Antigen-navn ADNP
Environmentally Preferable
Antigen synonymer HVDAS|MRD28|ADNP1
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Cy5®
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 23394
Isotype IgG
ImmunoKjemi Yes
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at 4°C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR