Anti-DFFA Rabbit Polyclonal Antibody

Leverandør: Bioss
BS-0214R
BOSSBS-0214REA 8500 NOK
BOSSBS-0214R
Anti-DFFA Rabbit Polyclonal Antibody
Antistoffer
Inhibitor of the caspase-activated DNase (DFF40).

IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: DFFA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR