Anti-ICAD Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Leverandør: Bioss
BS-0214R-A750
BOSSBS-0214R-A750EA 10425 NOK
BOSSBS-0214R-A750
Anti-ICAD Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antistoffer
Inhibitor of the caspase-activated DNase (DFF40).

Type: Primary
Antigen: ICAD
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 2-100/331
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR