Anti-DFFA Rabbit Polyclonal Antibody (Cy3®)

Leverandør: Bioss
BS-0214R-CY3
BOSSBS-0214R-CY3EA 10550 NOK
BOSSBS-0214R-CY3
Anti-DFFA Rabbit Polyclonal Antibody (Cy3®)
Antistoffer
Inhibitor of the caspase-activated DNase (DFF40).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DFFA
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR