Anti-GRIN2B Rabbit Polyclonal Antibody (Cy3®)

Leverandør: Bioss
BS-0222R-CY3
BOSSBS-0222R-CY3EA 10650 NOK
BOSSBS-0222R-CY3
Anti-GRIN2B Rabbit Polyclonal Antibody (Cy3®)
Antistoffer
NMDA receptor subtype of glutamate-gated ion channels with high calcium permeability and voltage-dependent sensitivity to magnesium. Mediated by glycine. In concert with DAPK1 at extrasynaptic sites, acts as a central mediator for stroke damage. Its phosphorylation at Ser-1303 by DAPK1 enhances synaptic NMDA receptor channel activity inducing injurious Ca2+ influx through them, resulting in an irreversible neuronal death (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NMDAR2B
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
More Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol GRIN2B
Antigen-navn Glutamate Ionotropic Receptor NMDA Type Subunit 2B
Environmentally Preferable
Antigen synonymer hNR3|GluN2B|NR2B|EIEE27|MRD6|NMDAR2B|glutamate ionotropic receptor NMDA type subunit 2B
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Cy3®
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 2904
Isotype IgG
ImmunoKjemi Yes
Immunogen 465-515/1482
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at 4°C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR