Anti-NAMPT Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Leverandør: Bioss
BS-0272R-A488
BOSSBS-0272R-A488EA 10450 NOK
BOSSBS-0272R-A488
Anti-NAMPT Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antistoffer
Catalyzes the condensation of nicotinamide with 5-phosphoribosyl-1-pyrophosphate to yield nicotinamide mononucleotide, an intermediate in the biosynthesis of NAD. It is the rate limiting component in the mammalian NAD biosynthesis pathway. The secreted form behaves both as a cytokine with immunomodulating properties and an adipokine with anti-diabetic properties, it has no enzymatic activity, partly because of lack of activation by ATP, which has a low level in extracellular space and plasma. Plays a role in the modulation of circadian clock function. NAMPT-dependent oscillatory productionMore Product Information