Anti-PBEF CT Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Leverandør: Bioss
BS-0273R-A350
BOSSBS-0273R-A350EA 10450 NOK
BOSSBS-0273R-A350
Anti-PBEF CT Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antistoffer
icotinamide phosphoribosyltransferase (NAmPRTase) (Nampt) (Pre-B cell-enhancing factor) (Pre-B-cell colony-enhancing factor 1) (Fisfatin) Catalyzes the condensation of nicotinamide with5-phosphoribosyl-1-pyrophosphate to yield nicotinamide mononucleotide, an intermediate in the biosynthesis of NAD. It is the rate limiting component in the mammalian NAD biosynthesis pathway. Expressed in large amounts in bone marrow, liver tissue, and muscle. Also present in heart, placenta, lung, and kidney tissues. Subcellular Location is in Cytoplasm. Belongs to the NAPRTase family.
RecommendedMore Product Information